missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cofilin Mouse anti-Human, Mouse, Rat, Clone: 1G6A2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66057-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Cofilin binds to F-actin and exhibits pH-sensitive F-actin depolymerizing activity. Regulates actin cytoskeleton dynamics. Important for normal progress through mitosis and normal cytokinesis. Plays a role in the regulation of cell morphology and cytoskeletal organization (By similarity). Required for the up-regulation of atypical chemokine receptor ACKR2 from endosomal compartment to cell membrane, increasing its efficiency in chemokine uptake and degradation. Required for neural tube morphogenesis and neural crest cell migration (By similarity).Specifica
Cofilin | |
Monoclonal | |
0.81 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P18760, P23528, P45592 | |
Cfl1 | |
Cofilin Fusion Protein Ag18044 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1G6A2 | |
Unconjugated | |
CFL1 | |
18 kDa phosphoprotein, CFL, CFL1, Cofilin, Cofilin 1, cofilin 1 (non muscle), Cofilin, non muscle isoform, p18 | |
Mouse | |
Protein A | |
RUO | |
1072, 12631, 29271 | |
-20°C | |
Liquid |