missing translation for 'onlineSavingsMsg'
Learn More

Cofilin Mouse anti-Human, Mouse, Rat, Clone: 1G6A2, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66057-1-IG-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897585

  • 140.00€ / 20µL
Spedizione stimata: 17-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Cofilin binds to F-actin and exhibits pH-sensitive F-actin depolymerizing activity. Regulates actin cytoskeleton dynamics. Important for normal progress through mitosis and normal cytokinesis. Plays a role in the regulation of cell morphology and cytoskeletal organization (By similarity). Required for the up-regulation of atypical chemokine receptor ACKR2 from endosomal compartment to cell membrane, increasing its efficiency in chemokine uptake and degradation. Required for neural tube morphogenesis and neural crest cell migration (By similarity).
Specifica

Specifica

Cofilin
Monoclonal
0.81 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P18760, P23528, P45592
Cfl1
Cofilin Fusion Protein Ag18044
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG2a
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence
1G6A2
Unconjugated
CFL1
18 kDa phosphoprotein, CFL, CFL1, Cofilin, Cofilin 1, cofilin 1 (non muscle), Cofilin, non muscle isoform, p18
Mouse
Protein A
RUO
1072, 12631, 29271
-20°C
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali