Learn More
Fisherbrand™ Combs for CHU25 Maxi-Plus Cooled Horizontal Gel Electrophoresis Unit
For use with CHU25 Maxi-Plus Cooled Horizontal Gel Electrophoresis Unit
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 2.00000kg
Descrizione
- Color coded
- Height adjustable
- Offer complete control over loading volume and well depth
Specifica
CHU25 Maxi-Plus Cooled Horizontal Gel Electrophoresis Unit | |
52 | |
20 μL |
Comb | |
1.5 mm | |
3 mm |