Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 0.75mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
- Recommended for ultra resolved bands
- 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifica
Black | |
OmniPAGE Mini Vertical Gel Electrophoresis Unit | |
12 | |
0.75 mm |
Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations | |
Comb | |
1 Each | |
28 μL |