Learn More
Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 0.75mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- For tightly resolved bands
- Recommended for high resolution gels and publication quality data
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 10-sample and 20-sample combs are Multichannel pipettor compatible
Specifica
Black | |
Comb | |
1 Each | |
118 μL |
Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit | |
4 Prep, 2 Marker | |
0.75 mm |