Learn More
Tenak™ Comfort Rack for Upright Freezers
Increases flexibility in the laboratory. Tenak™ Comfort Racks for Upright Freezers insure a more ergonomical operating process when working with samples.
Marca: Tenak™ TE24156
Dettagli aggiuntivi : Peso : 6.20000kg
Descrizione
- Cryoboxes placed in drawers
- Reduced operating weight
- Racks can be used with all freezer models
- Manufactured from stainless steel
- Sold without cryoboxes
Specifica
Comfort Rack | |
Boxes (40) | |
Upright Freezers | |
683 x 140 x 288mm | |
288 mm | |
683 mm |
8 x 5 | |
Stainless Steel | |
1 Pack | |
140 mm | |
32 mm |