Learn More
Lenz Laborglasintrumente Conical Joint Clips Type Keck™
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marca: Lenz Laborglasintrumente 01103745
Dettagli aggiuntivi : Peso : 0.60000kg
Descrizione
- For conical ground joints
- Material: POM, outstanding chemical resistance
- Operating temperature up to approx. 150 °C
Specifica
POM | |
For ground joint NS 45/40 | |
Brown |