missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPZ1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20440-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a subunit of the cytoplasmic coatamer protein complex, which is involved in autophagy and intracellular protein trafficking. The coatomer protein complex is comprised of seven subunits and functions as the coat protein of coat protein complex (COP)I-vesicles. Alternative splicing results in multiple transcript variants.Specifica
COPZ1 | |
Polyclonal | |
Unconjugated | |
COPZ1 | |
CGI 120, Coatomer subunit zeta 1, COPZ, COPZ1, HSPC181, Zeta 1 coat protein, Zeta 1 COP, zeta1 COP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22818, 315345, 56447 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P61923, P61924 | |
Copz1 | |
COPZ1 Fusion Protein Ag14234 | |
150 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |