missing translation for 'onlineSavingsMsg'
Learn More

COX11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11498-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898613

  • 140.00€ / 20µL
Spedizione stimata: 16-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Cytochrome c oxidase (COX) is the terminal enzyme in the electron transfer chain, functioning as a transmembrane proton pump that builds an electrochemical gradient with chemical energy from the reduction of O2. Cytochrome c oxidase assembly protein COX11 is an intracellular mitochondrial membrane protein necessary for the construction of an active COX complex. COX11 contains a single transmembrane helix downstream of the N-terminal, mitochondrial targeting sequence and a C-terminal Cu(I)-binding domain. The assembly of COX requires the delivery of metal cofactors. Along with COX12 and SCO1/2, COX11 acts as a metal ion chaperone necessary for copper insertion into CuA and CuB redox-active copper centers of COX in eukaryotes.
Specifica

Specifica

COX11
Polyclonal
Unconjugated
COX11
COX11, COX11P
Rabbit
Antigen Affinity Chromatography
RUO
1353, 690300, 69802
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6P8I6, Q9Y6N1
Cox11
COX11 Fusion Protein Ag2033
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali