missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11498-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Cytochrome c oxidase (COX) is the terminal enzyme in the electron transfer chain, functioning as a transmembrane proton pump that builds an electrochemical gradient with chemical energy from the reduction of O2. Cytochrome c oxidase assembly protein COX11 is an intracellular mitochondrial membrane protein necessary for the construction of an active COX complex. COX11 contains a single transmembrane helix downstream of the N-terminal, mitochondrial targeting sequence and a C-terminal Cu(I)-binding domain. The assembly of COX requires the delivery of metal cofactors. Along with COX12 and SCO1/2, COX11 acts as a metal ion chaperone necessary for copper insertion into CuA and CuB redox-active copper centers of COX in eukaryotes.Specifica
COX11 | |
Polyclonal | |
Unconjugated | |
COX11 | |
COX11, COX11P | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1353, 690300, 69802 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P8I6, Q9Y6N1 | |
Cox11 | |
COX11 Fusion Protein Ag2033 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |