missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX6A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11421-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Cytochrome c oxidase (COX), the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes polypeptide 2 (heart/muscle isoform) of subunit VIa, and polypeptide 2 is present only in striated muscles. Polypeptide 1 (liver isoform) of subunit VIa is encoded by a different gene, and is found in all non-muscle tissues. These two polypeptides share 66% amino acid sequence identity.Specifica
COX6A2 | |
Polyclonal | |
Unconjugated | |
COX6A2 | |
COX VIa M, COX6A, COX6A2, COX6AH, COXVIAH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12862, 1339, 25278 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P10817, P43023, Q02221 | |
COX6A2 | |
COX6A2 Fusion Protein Ag1985 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |