missing translation for 'onlineSavingsMsg'
Learn More

COX6A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11421-1-AP-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898483

  • 399.00€ / 150µL
Spedizione stimata: 17-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Cytochrome c oxidase (COX), the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes polypeptide 2 (heart/muscle isoform) of subunit VIa, and polypeptide 2 is present only in striated muscles. Polypeptide 1 (liver isoform) of subunit VIa is encoded by a different gene, and is found in all non-muscle tissues. These two polypeptides share 66% amino acid sequence identity.
Specifica

Specifica

COX6A2
Polyclonal
Unconjugated
COX6A2
COX VIa M, COX6A, COX6A2, COX6AH, COXVIAH
Rabbit
Antigen Affinity Chromatography
RUO
12862, 1339, 25278
-20°C
Liquid
Immunocytochemistry, Immunofluorescence
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P10817, P43023, Q02221
COX6A2
COX6A2 Fusion Protein Ag1985
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali