Learn More
JG Finneran Associates™ Tappi a capsula
Use these caps with standard 11mm chromatography vials. JG Finneran™ Crimp Caps feature an open top for filling and retrieving samples with a syringe.
Marca: JG Finneran Associates™ 5550-11
Dettagli aggiuntivi : Peso : 0.50000kg
Descrizione
- Made of the highest quality aluminum
- With PTFE seal
- Seals are available in a variety of colors for easy sample identification
Specifica
Silicone | |
PTFE, Silicone, Poly | |
1000 Pk. | |
11 mm |
Clear | |
PTFE/Silicone | |
Crimp Cap |