Learn More
JG Finneran Associates™ Tappi a capsula
Use these caps with standard 11mm chromatography vials. JG Finneran™ Crimp Caps feature an open top for filling and retrieving samples with a syringe.
Marca: JG Finneran Associates™ 5140-11Y
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Made of the highest quality aluminum
- With PTFE seal
- Seals are available in a variety of colors for easy sample identification
Specifica
Gomma | |
Alluminio, Rubber, Aluminum | |
Confezione da 1000 | |
11 mm |
Rosso, Giallo | |
PTFE trasparente/Gomma rossa | |
Tappo a capsula |