Learn More
Notrax Crossrib Sof-Tred™ Dyna-Shield™ Layer with Side-to-Side Wide Ribs, Black/Yellow
Notrax Crossrib Sof-Tred™ - Exceptional worker comfort due to 12.7 mm thick specially formulated compound. Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl.
Marca: Notrax 406S0035BY
Dettagli aggiuntivi : Peso : 5.30000kg
Descrizione
- Exceptional worker comfort due to 127 mm thick specially formulated compound
- Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl supported by independent laboratory testing
- Side-to-side wide ribs provide traction and easy to sweep clean
- Available with yellow borders along sides to comply with OSHA code 1910-144
- All 4 sides are bevelled and sealed to minimize tripping hazards
Specifica
59 in. | |
Industrial and Commercial Matting | |
91 cm |
Black/Yellow | |
Microcellular Vinyl | |
150 cm |