missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CX3CR1 Rat anti-Human , BB515, Clone: 2A9-1, BD Biosciences™
Rat Monoclonal Antibody
Marca: BD Biosciences 565903
Dettagli aggiuntivi : Peso : 0.00060kg
Beschreibung
CX3CR1 Monoclonal antibody specifically detects CX3CR1 in Human samples. It is validated for Flow CytometrySpezifikation
CX3CR1 | |
Monoclonal | |
Brilliant Blue 515 (BB515) | |
CCRL1; CMKBRL1; CMKDR1; GPR13; GPRV28; V28; Fractalkine Receptor | |
Human CX3CR1 Recombinant Protein | |
RUO | |
Human | |
IgG2b κ |
Flow Cytometry | |
2A9-1 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. |