Learn More
ACROBiosystems Cynomolgus CD24 Protein, Fc Tag
Marca: ACROBiosystems CD4-C5257-100ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
CD24 may have a pivotal role in cell differentiation of different cell types. Signaling could be triggered by the binding of a lectin-like ligand to the CD24 carbohydrates, and transduced by the release of second messengers derived from the GPI-anchor. Modulates B-cell activation responses. Promotes AG-dependent proliferation of B-cells, and prevents their terminal differentiation into antibody-forming cells. In association with SIGLEC10 may be involved in the selective suppression of the immune response to danger-associated molecular patterns (DAMPs) such as HMGB1, HSP70 and HSP90. Plays a role in the control of autoimmunity.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 29.3 kDa. The protein migrates as 42-50 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.Specifica
29.3 kDa | |
Cynomolgus CD24, Fc Tag (CD4-C5257) is expressed from human 293 cells (HEK293). It contains AA Thr 27 - Lys 54 (Accession # I7GKK1-1). | |
-20°C | |
CD24 | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
100 μg | |
Thr 27 - Lys 54 | |
CD24,CD24A | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |