Learn More
Cytokeratin 17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18502-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes the type I intermediate filament chain keratin 17, expressed in nail bed, hair follicle, sebaceous glands, and other epidermal appendages. Mutations in this gene lead to Jackson-Lawler type pachyonychia congenita and steatocystoma multiplex.Specifica
Cytokeratin 17 | |
Polyclonal | |
Unconjugated | |
KRT17 | |
39.1, CK 17, Cytokeratin 17, K17, keratin 17, KRT17, PC, PC2, PCHC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16667, 287702, 3872 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Flow Cytometry | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q04695, Q6IFU8, Q9QWL7 | |
Krt17 | |
Cytokeratin 17 Fusion Protein Ag13351 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |