Learn More
Trajan Scientific and Medical™ Deactivated (Methyl) Fused Silica Tubing
Trajan methyl deactivated fused silica capillary tubing is an essential ingredient in maintaining a high performance chromatography system.
Marca:  Trajan Scientific and Medical™ 0624491
Dettagli aggiuntivi : Peso : 0.01000kg
Codice prodotto. 15208823
  • 41.65€ / Pezzo
Spedizione stimata: 10-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Trajan methyl deactivated fused silica capillary tubing is an essential ingredient in maintaining a high performance chromatography system.
  • Chemically inert and thermally stable up to 380°C
  • Suitable for organic and aqueous solvents
  • Ideal for biotechnology applications
Specifica
Specifica
Organic and Aqueous Solvents
5/Pk.
300 μm
Thermally stable up to 380°C
Tubing
170 μm
2 m