Learn More
Trajan Scientific and Medical™ Deactivated (Methyl) Fused Silica Tubing
Trajan methyl deactivated fused silica capillary tubing is an essential ingredient in maintaining a high performance chromatography system.
Marca:  Trajan Scientific and Medical™ 062444
Dettagli aggiuntivi : Peso : 0.01000kg
Codice prodotto. 15218803
  • 41.65€ / 2m
Spedizione stimata: 05-07-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Trajan methyl deactivated fused silica capillary tubing is an essential ingredient in maintaining a high performance chromatography system.
  • Chemically inert and thermally stable up to 380°C
  • Suitable for organic and aqueous solvents
  • Ideal for biotechnology applications
Specifica
Specifica
Organic and Aqueous Solvents
1/Pk.
220 μm
Thermally stable up to 380°C
Tubing
220 μm
2 m