Learn More
Digi-Sense™ Borescopes
Cole Parmer Digi-Sense Borescopes features a flexible, camera-tipped probe. Ideal for inspecting duct work, wiring locations, piping and vehicles.
Marca: Digi-Sense™ 20250-27
Dettagli aggiuntivi : Peso : 2.18200kg Transport : UN number : 3481 Chem class : 9
Includes: Probe, Mirror, Hook, Magnet, Carrying case, USB cable, DC 5V charger, Video output cable, Rechargable 3.7V lithium battery
Descrizione
- Camera-tipped probe provides easy viewing in hard-to-reach areas
- Bright LCD monitor allows instant viewing of area to be inspected
- Adjustable LED light on camera tip provides the necessary light in dimly lit areas or adjusts down when viewing reflective objects to eliminate washout
Compliance
CE; IP67
Servizi
2 year
Specifica
Color LCD | |
CE; IP67 | |
IP67 | |
4 adjustable LED lights | |
3.2 in. | |
Yes |
Built-in lithium battery | |
8 GB flash memory | |
English, French, German, Spanish | |
Battery; USB charging | |
Still and video images for playback |