missing translation for 'onlineSavingsMsg'
Learn More

DNA polymerase beta Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 18003-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898024

  • 129.00€ / 20µL

Spedizione stimata: 28-12-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

DNA polymerase beta comprises an amino-terminal 8-kDa domain and a carboxy-terminal 31-kDa domain. The N-terminal ssDNA binding domain has a deoxyribose phosphodiesterase activity while the C-terminal domain has a nucleotidyltransferase activity. Mammalian DNA polymerase beta, a DNA repair polymerase, is constitutively expressed in cultured cells, but treatment of cells with the DNA-alkylating agents such as N-methyl-N′-nitro-N-nitrosoguanidine (MNNG) or methyl methanesulfonate up-regulates beta-pol level. DNA polymerase beta fills single nucleotide gaps in DNA produced by the base excision repair pathway of mammalian cells.
Specifica

Specifica

DNA polymerase beta
Polyclonal
Unconjugated
POLB
DNA polymerase beta, POLB
Rabbit
Antigen Affinity Chromatography
RUO
18970, 29240, 5423
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P06746, P06766, Q8K409
Polb
DNA polymerase beta Fusion Protein Ag12496
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali