missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA polymerase beta Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18003-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
DNA polymerase beta comprises an amino-terminal 8-kDa domain and a carboxy-terminal 31-kDa domain. The N-terminal ssDNA binding domain has a deoxyribose phosphodiesterase activity while the C-terminal domain has a nucleotidyltransferase activity. Mammalian DNA polymerase beta, a DNA repair polymerase, is constitutively expressed in cultured cells, but treatment of cells with the DNA-alkylating agents such as N-methyl-N′-nitro-N-nitrosoguanidine (MNNG) or methyl methanesulfonate up-regulates beta-pol level. DNA polymerase beta fills single nucleotide gaps in DNA produced by the base excision repair pathway of mammalian cells.Specifica
DNA polymerase beta | |
Polyclonal | |
Unconjugated | |
POLB | |
DNA polymerase beta, POLB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18970, 29240, 5423 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P06746, P06766, Q8K409 | |
Polb | |
DNA polymerase beta Fusion Protein Ag12496 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |