missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12096-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This antibody is specific to DNAJC19 and had been tested by siRNA.
The protein encoded by this gene is thought to be part of a complex involved in the ATP-dependent transport of transit peptide-containing proteins from the inner cell membrane to the mitochondrial matrix. Defects in this gene are a cause of 3-methylglutaconic aciduria type 5 (MGA5), also known as dilated cardiomyopathy with ataxia (DCMA). Alternative splicing of this gene results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 1, 2, 6, 10, 14 and 19.Specifica
DNAJC19 | |
Polyclonal | |
Unconjugated | |
DNAJC19 | |
DNAJC19, TIM14, TIMM14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
131118, 502525, 67713 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96DA6, Q9CQV7 | |
Dnajc19 | |
DNAJC19 Fusion Protein Ag2739 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |