Learn More
Dräger™ CMS Chips: Acetone
Suitable for 10 measurements of one gas
Marca: Dräger™ 6406470
Dettagli aggiuntivi : Peso : 0.02880kg
Descrizione
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.Specifica
700 to 1100 hpa | |
0 to 30mg/L | |
60 to 600 sec. | |
5° to 40°C |
Acetone CMS Chip | |
40 to 600 | |
41°F to 104°F |