missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DTX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18565-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
DTX2 functions as an E3 ubiquitin ligase. Regulator of Notch signaling, a signaling pathway involved in cell-cell communications that regulates a broad spectrum of cell-fate determinations. Probably acts both as a positive and negative regulator of Notch, depending on the developmental and cell context. Mediates the antineural activity of Notch, possibly by inhibiting the transcriptional activation mediated by MATCH1. Functions as an ubiquitin ligase protein in vitro, suggesting that it may regulate the Notch pathway via some ubiquitin ligase activity.Specifica
DTX2 | |
Polyclonal | |
Unconjugated | |
DTX2 | |
deltex homolog 2 (Drosophila), Deltex2, DTX2, hDTX2, KIAA1528, Protein deltex 2, RING finger protein 58, RNF58 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
113878, 304591, 74198 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q86UW9, Q8R3P2 | |
Dtx2 | |
DTX2 Fusion Protein Ag4376 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |