missing translation for 'onlineSavingsMsg'
Learn More

DTX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 18565-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898534

  • 140.00€ / 20µL
Spedizione stimata: 22-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

DTX2 functions as an E3 ubiquitin ligase. Regulator of Notch signaling, a signaling pathway involved in cell-cell communications that regulates a broad spectrum of cell-fate determinations. Probably acts both as a positive and negative regulator of Notch, depending on the developmental and cell context. Mediates the antineural activity of Notch, possibly by inhibiting the transcriptional activation mediated by MATCH1. Functions as an ubiquitin ligase protein in vitro, suggesting that it may regulate the Notch pathway via some ubiquitin ligase activity.
Specifica

Specifica

DTX2
Polyclonal
Unconjugated
DTX2
deltex homolog 2 (Drosophila), Deltex2, DTX2, hDTX2, KIAA1528, Protein deltex 2, RING finger protein 58, RNF58
Rabbit
Antigen Affinity Chromatography
RUO
113878, 304591, 74198
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q86UW9, Q8R3P2
Dtx2
DTX2 Fusion Protein Ag4376
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali