Learn More
DWK Life Sciences DURAN™ Funnel, ribbed
A funnel enables a clean transfer and filtration of liquids.
Marca: DWK Life Sciences 213526105
73.20 EUR valido fino al 2024-12-31
Usa il codice promo "23258" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.51000kg
Descrizione
Transferring liquids and solids is a common working method. In this activity the funnel is used, which due to its shape can absorb a relatively large amount of liquid or solid and transfer it through a constriction into a vessel with a small opening. Funnels are available in different sizes and designs, selected according to their application.
- A sticking to a smooth glass surface and thus a slowing down of the filtration speed is prevented by the ribbed shape
- The ribbed form is ideal for filtering with round-paper
- Fired-on, highly durable white print
Specifica
200 mm | |
Yes | |
1 Ea. |
Boro 3.3 | |
Filtering of liquids; The spillage of substances is avoided; Filtering |