Learn More
DWK Life Sciences DURAN™ Glass Stopper, Clear, Semi-hollow, Standard Taper Ground Joint, Octagonal flat head
Clear DURAN™ glass ground stopper.
Marca:  DWK Life Sciences 216241603
52.01 EUR valido fino al 2024-12-31
Usa il codice promo "23258" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.51500kg
Codice prodotto. 10010141
  • 54.75€ / Pezzo
Spedizione stimata: 18-07-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Semi-hollow or solid DURAN™ glass stoppers can be used to close glassware items such as flasks, separatory funnels or reagent bottles with ground joint necks. The Standard Taper ground glass joint forms a tight, leak-proof seal. Use grease for heating or vacuum applications. With octagonal shaped head. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!
  • All DURAN™ glass components and therefore suitable for storage of aggressive materials
  • Tight closure due to Standard Taper ground joint
  • Octagonal shaped head
Specifica
Specifica
Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders
Boro 3.3
No
1 Ea.
ST 85/55