Learn More
DWK Life Sciences DURAN™ Plain Cylinder, with round base, without graduation
Cylinders without graduation can be used to mix components.
Marca: DWK Life Sciences 213994609
304.00 EUR valido fino al 2024-12-31
Usa il codice promo "23258" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 3.60000kg
Descrizione
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility.
- Plain ground rim
- With round base
Specifica
Borosilicate Glass (3.3) | |
550 mL | |
10/Pk. |
60 mm | |
Diluting solutions; Mixing several components | |
Yes |