Learn More
DWK Life Sciences DURAN™ Stopcock Key, hollow, threaded glass handle, ground, with three part retaining device
To be used for higher tightness requirements.
Marca: DWK Life Sciences 286047009
1249.25 EUR valido fino al 2024-12-31
Usa il codice promo "23258" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 2.50000kg
Descrizione
Hollow DURAN™ tap plugs are well suited for work with temperature changes and higher tightness requirements.
- Hollow tap plug with thread, glass handle and 3-piece tap safety device
Specifica
8 mm | |
Yes | |
25/Pk. |
Borosilicate Glass (3.3), Polypropylene | |
24 NS | |
When working with temperature changes and higher tightness requirements |