Learn More
Lenz Laborglasinstrumente™ Duran Tubing Reaction Vessels
Marca: Lenz Laborglasinstrumente™ 06001158
Dettagli aggiuntivi : Peso : 1.00000kg
Specifica
500 mL | |
220 mm | |
Cylindrical |
Borosilicate Glass, Duran Glass | |
Reaction vessel | |
Round |