Learn More
Lenz Laborglasinstrumente™ Duran Tubing Reaction Vessels
Marca: Lenz Laborglasinstrumente™ 06000488
Dettagli aggiuntivi : Peso : 1.00000kg
Specifica
10 L | |
410 mm | |
Cylindrical |
Borosilicate Glass, Duran Glass | |
Reaction vessel | |
Flat |