missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dymeclin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12001-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a protein which is necessary for normal skeletal development and brain function. Mutations in this gene are associated with two types of recessive osteochondrodysplasia, Dyggve-Melchior-Clausen (DMC) dysplasia and Smith-McCort (SMC) dysplasia, which involve both skeletal defects and mental retardation.Specifica
Dymeclin | |
Polyclonal | |
Unconjugated | |
DYM | |
DMC, DYM, Dymeclin, SMC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
291433, 54808, 69190 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
B4F766, Q7RTS9, Q8CHY3 | |
Dym | |
Dymeclin Fusion Protein Ag2600 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |