Learn More
Trajan™ EC GC Septa
EC GC septa is designed for long lifetime and low injection port adhesion
Marca: Trajan™ 041843
Dettagli aggiuntivi : Peso : 0.03600kg
Descrizione
- Combines long injection lifetime, low bleed and low injection port adhesion
- Reliable seal against carrier gas pressure in the inlet
- No bleed or contamination into the GC system
- Maximum operating temperature of 350°C (300°C for 17mm size)
Specifica
8 mm | |
350°C | |
50/Pk. |
High Temperature Silicon | |
Long Life EC Septa |