Learn More
Trajan™ EC GC Septa
EC GC septa is designed for long lifetime and low injection port adhesion
Marca: Trajan™ 041903
Dettagli aggiuntivi : Peso : 0.50000kg
Descrizione
- Combines long injection lifetime, low bleed and low injection port adhesion
- Reliable seal against carrier gas pressure in the inlet
- No bleed or contamination into the GC system
- Maximum operating temperature of 350°C (300°C for 17mm size)
Specifica
17 mm | |
300°C | |
Long Life EC Septum |
High Temperature Silicone | |
Yes | |
24/Pk. |