missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF3J Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10439-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Eukaryotic initiation factor-3 (EIF3) has a molecular mass of about 600 kD and contains 13 nonidentical protein subunits, including EIF3J. EIF3 plays a central role in binding of initiator methionyl-tRNA and mRNA to the 40S ribosomal subunit to form the 40S initiation complex (Fraser et al., 2004; Fraser et al., 2007).Specifica
EIF3J | |
Polyclonal | |
Unconjugated | |
EIF3J | |
eIF 3 alpha, eIF3 alpha, eIF3 p35, EIF3J, EIF3S1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
691947, 78655, 8669 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A0JPM9, O75822, Q3UGC7 | |
EIF3J, Eif3j1 | |
EIF3J Fusion Protein Ag0678 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |