Learn More
Merck Piastre filtro HTS a 96 pozzetti MultiScreen™ con membrana MCE
Commonly used for Elispot assays, bacterial screening, ELISA or viral infection studies
Marca: Merck MSHAS4510
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- Developed for high-throughput use with automated work stations
- Individually sealed wells prevent incubation crosstalk
- Removable underdrain allows access to the filters
- Rigid sidewalls improve handling
- Ample surfaces provided for bar code labels
- Pore size: .45μm
Solid-phase RIA, DNA hybridization, ELISA and protein binding
Specifica
Standard | |
8 x 12 | |
Trasparente | |
96 | |
0.28 cm2 | |
10/conf. |
Esteri misti di cellulosa | |
0.45 μm | |
Stirene | |
Sterile | |
RIA fase solida, ibridazione del DNA, ELISA, legame delle proteine |