Learn More
Merck MultiScreen™ Multiple Punch and Punch Tips
For rdionucleotide counting by transfer of individual filters to tubes
Marca: Merck MAMP09608
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Punch and tips are used to simultaneously remove filters from eight wells of a MultiScreen plate and distribute them into eight scintillation vials for scintillation or gamma radioscope detection
- Remove filters from wells separately, eliminating cross-contamination
- Usable with all MultiScreen plates containing Durapore (PVDF) membrane; mixed cellulose ester membranes (HA) and glass fiber membranes must be wet when punched from plate
- Multiple punch includes punch with punch carrier plate and punch distributor
Order Info
Disposable punch tips for the multiple punch are available. Three punch kits are available—all include multiple punch and carrier racks
Specifica
Punch kit | |
Radionucleotide counting by transfer of individual filters to tubes | |
Punch with punch carrier plate and punch distributor | |
1/Pk. |