Learn More
Merck Samplicity™ Filtration System Vial Trays
Vacuum filtration system for sample preparation for HPLC
Marca: Merck SAMWASTTR
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Overview of Samplicity™ G2 System- Samplicity™ G2 Filtration System enables vacuum driven filtration of up to 8 samples at once, directly into HPLC vials.
- Filtration for HPLC sample prep without syringes. Sample is added to an adapter funnel on a standard Millex™ 33 mm filter (Millex™ Filters for the Samplicity™ G2 System) or an integrated funnel and filter (Millex Samplicity™ filter)
- Samplicity™ G2 System has lower ergonomic impact than manual syringe filtration
Applications: HPLC; Filtration of viscous or particulate laden samples for HPLC
Specifica
Samplicity G2 Filtration System | |
Non-sterile | |
5 Trays |
Recycled Polyester | |
Analytical Sample Prep, High Performance Liquid Chromatography (HPLC) | |
Waste Trays |