Learn More
Merck Parti di ricambio per sistema filtro a vuoto asettico Sterifil™ 47 mm
Used for general filtration applications and filtration of samples for particulate or biological contamination analysis
Marca: Merck XX1104703
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
MilliporeSigma Polypropylene Support Screen, For Use With: For Swinnex 47, Surface Area: Support Screen, 47mm dia. Filter, Polypropylene, for Swinnex 47
For use with Sterifil™ Aseptic SystemsSpecifica
Not Autoclavable | |
Sterifil aseptic system | |
Non-sterile |
Funnel Cover | |
Polysulfone Funnel Cover | |
Laboratory Filter Holders or Cyclones |