missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETV5 Mouse anti-Human, Clone: 2A4A4, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66657-1-IG-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
ETS variant 5,transcription factor of the ETS family,divergent member of the winged helix-turn-helix superfamily,expressed in brain,placenta and other tissues involved in development. Tissue specificity: Ubiquitous. ERM is expressed in Sertoli cells and spermatogonial stem cells (SSCs). Studies on knockout mice have shown that it is required for SSC self-renewal but not differentiation, thus affected mice exhibit progressive germ cell depletion.Specifica
ETV5 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P41161 | |
ETV5 | |
ETV5 Fusion Protein Ag3708 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot | |
2A4A4 | |
Unconjugated | |
ETV5 | |
ERM, Ets related protein ERM, ETS translocation variant 5, ets variant 5, ETV5 | |
Mouse | |
Protein A | |
RUO | |
2119 | |
-20°C | |
Liquid |