missing translation for 'onlineSavingsMsg'
Learn More

ETV5 Mouse anti-Human, Clone: 2A4A4, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66657-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898735

  • 325.38€ / 150µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

ETS variant 5,transcription factor of the ETS family,divergent member of the winged helix-turn-helix superfamily,expressed in brain,placenta and other tissues involved in development. Tissue specificity: Ubiquitous. ERM is expressed in Sertoli cells and spermatogonial stem cells (SSCs). Studies on knockout mice have shown that it is required for SSC self-renewal but not differentiation, thus affected mice exhibit progressive germ cell depletion.
Specifica

Specifica

ETV5
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P41161
ETV5
ETV5 Fusion Protein Ag3708
150 μL
Primary
Human
Antibody
IgG2a
Western Blot
2A4A4
Unconjugated
ETV5
ERM, Ets related protein ERM, ETS translocation variant 5, ets variant 5, ETV5
Mouse
Protein A
RUO
2119
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali