Learn More
Thermo Scientific™ EZ-Link™ NHS-PEG12-biotina
Biotinylate protein amines and maximize solubility of proteins with this very long (56.0A) NHS-ester biotinylation reagent containing a pegylated, water-soluble spacer arm.
Marca: Thermo Scientific™ 21313
907.44 EUR valido fino al 2024-06-28
Usa il codice promo "21649" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
Thermo Scientific™ EZ-Link NHS-PEG12-Biotin is very long, pegylated biotinylation reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.
Highlights:
- Water-soluble polyethylene glycol (PEG) spacer arm increases solubility of labeled protein
- Excellent purity, molecular weight uniformity and amine-reactivity
- Long spacer arm reduces steric hindrance when binding to avidin molecules
Specifica
NHS-PEG12-Biotin | |
Store desiccated at -20°C. Shipped at ambient temperature. | |
Cell-Impermeant | |
Active Ester, Succinimidyl Ester, NHS Ester | |
Biotin | |
Biotin & Analogs |
500 mg | |
EZ-Link™ | |
Amine | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water | |
Extra-long, Pegylated |
For Research Use Only. Not for use in diagnostic procedures.