missing translation for 'onlineSavingsMsg'
Learn More

FCGR2A / CD32a Mouse anti-Human, Clone: 1D3F1, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66529-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898485

  • 325.38€ / 150µL

Spedizione stimata: 06-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
Specifica

Specifica

FCGR2A / CD32a
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P12318
FCGR2A
FCGR2A / CD32a Fusion Protein Ag25878
150 μL
Primary
Human
Antibody
IgG2b
Western Blot, Immunohistochemistry (Paraffin)
1D3F1
Unconjugated
FCGR2A
CD32, CD32A, CDw32, Fc gamma RII a, Fc gamma RIIa, FCG2, FcGR, FCGR2, FCGR2A, FCGR2A1, FcRII a, IGFR2, IgG Fc receptor II a
Mouse
Protein A
RUO
2212
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali