missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11028-2-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This antibody recognizes specifically the 34 kDa FHL3 protein.
LIM proteins are defined by the possession of a highly conserved double zinc finger motif called the LIM domain.Specifica
FHL3 | |
Polyclonal | |
Unconjugated | |
FHL3 | |
FHL 3, FHL3, four and a half LIM domains 3, Skeletal muscle LIM protein 2, SLIM 2, SLIM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14201, 2275, 313582 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13643, Q9R059 | |
Fhl3 | |
FHL3 Fusion Protein Ag1495 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |