Learn More
Hamilton™ Filtered Low Retention Pre-Sterilized Racked Pipette Tips
With surface walls completely free of occlusions and cavities so fluid retention is virtually eliminated.
Marca:  Hamilton™ 68007-23
Dettagli aggiuntivi : Peso : 0.20000kg
Codice prodotto. 15335094
  • 213.00€ / pezzi 10
Spedizione stimata: 10-07-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Manufactured from chemically resistant virgin polypropylene for excellent clarity and smooth surfaces
  • Pre-sterilized
  • 96 tips to a rack
  • Delivering outstanding accuracy and precision, guaranteed
Specifica
Specifica
10 μL
Sterile
Polypropylene
Filtered
Rack