Learn More
Fisherbrand™ Combs for Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis, 1mm Thick
For use with Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis units.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- Recommended for high resolution gels and publication quality data
- Supplied as standard
- 18-sample and 36-sample combs are Multichannel pipettor compatible
Specifica
White | |
Comb | |
1 Each | |
25 μL |
Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis Units | |
36 | |
1 mm |