Learn More
Fisherbrand™ Sacchetti per campioni a tenuta “Zipper”
La chiusura a zip consente di sigillare le buste
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
La chiusura a zip consente di sigillare le buste
- Durable, transparent polyethylene
- Ideal for collecting, storing, and protecting laboratory specimens and samples easily and conveniently
- Clear, print-free surface lets user mark and label each bag separately
Not suggested for use with liquids
Specifica
Not Autoclavable | |
Zip Seal | |
LDPE | |
50 | |
13 x 9 in., (33 x 23cm) | |
33 cm | |
23 cm |
3.25 lb. | |
Clear | |
250 | |
4 mil | |
13 in. | |
9 in. |