missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20268-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The antibody is specific to FOXH1.
FOXH1 is a member of the Forkhead-box (FOX) family of transcription factors and is the human homolog of Xenopus forkhead activin signal transducer-1. FOXH1 plays an important role in mediating TGF-beta/activin signals through its interaction with the SMAD2-SMAD4 complex. It is expressed primarily during early development and localizes in the cytoplasm until it forms a complex with the SMAD proteins at which time it translocates to the nucleus to positively or negatively affect gene expression.Specifica
FOXH1 | |
Polyclonal | |
Unconjugated | |
FOXH1 | |
FAST 1, Fast 2, FAST1, FAST2, forkhead box H1, Forkhead box protein H1, FOXH1, hFAST 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14106, 300054, 8928 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75593, O88621 | |
FOXH1 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |